Trimethylsilanol
Trimethylsilanol
Trimethylsilanol
Thermo Scientific Chemicals

Trimethylsilanol

CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol
Quantity:
5 g
25 g
100 g
Catalog number 458571000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1066-40-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only