Mycophenolate mofetil, 98%
Mycophenolate mofetil, 98%
Mycophenolate mofetil, 98%
Thermo Scientific Chemicals

Mycophenolate mofetil, 98%

CAS: 128794-94-5 | C23H31NO7 | 433.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459370250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS128794-94-5
IUPAC Name2-(morpholin-4-yl)ethyl (4E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydro-2-benzofuran-5-yl)-4-methylhex-4-enoate
Molecular FormulaC23H31NO7
InChI KeyRTGDFNSFWBGLEC-SYZQJQIISA-N
SMILESCOC1=C(C)C2=C(C(=O)OC2)C(O)=C1C\C=C(/C)CCC(=O)OCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point93°C to 98°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only