CD1d Monoclonal Antibody (51.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD1d Monoclonal Antibody (51.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD1d Monoclonal Antibody (51.1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0016-41, 46-0016-42)