TCR V beta 12 Monoclonal Antibody (MR11-1), PerCP-eFluor™ 710, eBioscience™ - FAQs
TCR V beta 12 Monoclonal Antibody (MR11-1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TCR V beta 12 Monoclonal Antibody (MR11-1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5798-80)