VISTA Monoclonal Antibody (MH5A), PerCP-eFluor™ 710, eBioscience™ - FAQs
VISTA Monoclonal Antibody (MH5A), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for VISTA Monoclonal Antibody (MH5A), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-5919-80, 46-5919-82)