CD369 (Clec7a, Dectin-1) Monoclonal Antibody (15E2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD369 (Clec7a, Dectin-1) Monoclonal Antibody (15E2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD369 (Clec7a, Dectin-1) Monoclonal Antibody (15E2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9856-42, 46-9856-41)