Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Thermo Scientific Chemicals

Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination

CAS: 14552-35-3 | C4H18CuN4O2 | 217.76 g/mol
Have Questions?
Catalog NumberQuantity
4606800101 L
Catalog number 460680010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS14552-35-3
IUPAC Namecopper(2+) bis(ethane-1,2-diamine) dihydroxide
Molecular FormulaC4H18CuN4O2
InChI KeyWATCRQGYOIZIHC-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Cu++].NCCN.NCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Strength0.98 to 1.02 M (copper molarity)
Appearance (Color)Dark blue
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only