Pyrantel pamoate, 98%
Pyrantel pamoate, 98%
Pyrantel pamoate, 98%
Thermo Scientific Chemicals

Pyrantel pamoate, 98%

CAS: 22204-24-6 | C11H14N2S·C23H16O6 | 594.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461890250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS22204-24-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Loss on drying=<2 % (60°C, 3 h) (vacuum)
Titration with HClO497.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only