Loxapine succinate
Thermo Scientific Chemicals

Loxapine succinate

CAS: 27833-64-3 | C18H18ClN3O·C4H6O4
Have Questions?
Catalog NumberQuantity
4626900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462690010
Price (USD)
292.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
292.00
Each
Chemical Identifiers
CAS27833-64-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White
Infrared spectrumConforms
Melting point150°C to 154°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only