Betulinic acid
Betulinic acid
Betulinic acid
Thermo Scientific Chemicals

Betulinic acid

CAS: 472-15-1 | C30H48O3 | 456.71 g/mol
製品番号(カタログ番号) 462870010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS472-15-1
IUPAC Name(1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysene-3a-carboxylic acid
Molecular FormulaC30H48O3
InChI KeyQGJZLNKBHJESQX-FZFNOLFKSA-N
SMILESCC(=C)[C@@H]1CC[C@@]2(CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12)C(O)=O
さらに表示
Appearance (Color)White to light beige
Loss on drying=<5 %
Appearance (Form)Powder
HPLC>=96.0 %
Solubility(0.1% in ethanol) Clear colorless
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only