Product Image
Thermo Scientific Chemicals

Scutellarin

CAS: 27740-01-8 | C21H18O12 | 462.36 g/mol
製品番号(カタログ番号) 465132500
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS27740-01-8
IUPAC Name(2S,3S,4S,5R,6S)-6-{[5,6-dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular FormulaC21H18O12
InChI KeyDJSISFGPUUYILV-ZFORQUDYSA-N
SMILESO[C@@H]1[C@@H](O)[C@H](OC2=C(O)C(O)=C3C(=O)C=C(OC3=C2)C2=CC=C(O)C=C2)O[C@@H]([C@H]1O)C(O)=O
さらに表示
Appearance (Color)Yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only