1-Methoxy-2-butanol, Thermo Scientific Chemicals
1-Methoxy-2-butanol, Thermo Scientific Chemicals
1-Methoxy-2-butanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methoxy-2-butanol, Thermo Scientific Chemicals

製品番号(カタログ番号) 466370250
価格(JPY)
-
数量:
25 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS53778-73-7
IUPAC Name1-methoxybutan-2-ol
Molecular FormulaC5H12O2
InChI KeyCSZZMFWKAQEMPB-UHFFFAOYNA-N
SMILESCCC(O)COC
さらに表示
Appearance (Color)Colorless
Infrared spectrumConforms
Refractive index1.4090 to 1.4130 (20°C, 589 nm)
GC>=93.0 %
Appearance (Form)liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only