Pierce™ Immunostain Enhancer
Thermo Scientific™

Pierce™ Immunostain Enhancer

Thermo Scientific Pierce Immunostain Enhancer enables significant antibody dilution (5- to 20-fold beyond normal) to increase signal intensity 3- toRead more
Have Questions?
Catalog NumberQuantity
4664420 mL
Catalog number 46644
Price (USD)
509.65
Online exclusive
548.00
Save 38.35
Each
Quantity:
20 mL
Request bulk or custom format
Price (USD)
509.65
Online exclusive
548.00
Save 38.35
Each
Thermo Scientific Pierce Immunostain Enhancer enables significant antibody dilution (5- to 20-fold beyond normal) to increase signal intensity 3- to 12-fold in immunostaining applications with chromogenic or fluorescent substrates.

Features of Immunostain Enhancer:

• Save precious antibody—Pierce Immunostain Enhancer allows the customer to use only a fraction of antibody to achieve the same signal as with conventional immunodetection
• Convenience—simply replace your current antibody dilution buffer with Pierce Immunostain Enhancer (unlike other signal enhancement methods which require additional steps)
• Increased signal intensity and sensitivity—provides 3- to 12- fold increase in signal intensity and sensitivity for improved visualization of the antigen of interest in cells and tissues
• Improved specificity—significantly improves signal-to-noise ratio for poor quality and low affinity antibodies
• Compatible—can be used with chromogenic and fluorescent detection methods

This ready-to-use enhancer for immunohistochemical staining does not add steps to the workflow because it is used to dilute the primary and secondary antibodies. The Pierce Immunostain Enhancer is compatible with fluorescence and chromogenic detection and routinely increases both signal intensity and detection sensitivity. Signal enhancement is antibody-dependent and typically ranges from 3- to 12-fold. Because of the strong signal enhancement, the Pierce Immunostain Enhancer reduces the amount of antibody required to achieve optimal detection.

The Pierce Immunostain Enhancer enables significant dilution of primary and secondary antibodies. Dilution 5- to 20-fold beyond the antibody vendor's recommendation has been used successfully; however, the dilution factor varies depending on the antibody, tissue, and antigen.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Immunohistochemistry
FormatLiquid
Quantity20 mL
Reagent TypeSignal Enhancers
Product LinePierce
TypeImmunostain Enhancer
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.