Dorzolamide hydrochloride
Thermo Scientific Chemicals

Dorzolamide hydrochloride

製品番号(カタログ番号) 466610010
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS130693-82-2
IUPAC Namehydrogen (2R,4R)-4-(ethylamino)-2-methyl-1,1-dioxo-2H,3H,4H-1λ⁶-thieno[2,3-b]thiopyran-6-sulfonamide chloride
Molecular FormulaC10H17ClN2O4S3
InChI KeyOSRUSFPMRGDLAG-CIRBGYJCSA-N
SMILES[H+].[Cl-].CCN[C@@H]1C[C@@H](C)S(=O)(=O)C2=C1C=C(S2)S(N)(=O)=O
さらに表示
Appearance (Color)White to almost white
Loss on drying=<0.5 % (1 g, 105°C)
Infrared spectrumConforms
HPLC>=98.0 %
Appearance (Form)crystalline powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only