Amprenavir
Thermo Scientific Chemicals

Amprenavir

CAS: 161814-49-9 | C25H35N3O6S, C25H35N3O6S | 505.63 g/mol
製品番号(カタログ番号) 466980250
価格(JPY)
-
数量:
25 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS161814-49-9
IUPAC Name(3S)-oxolan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate
Molecular FormulaC25H35N3O6S
InChI KeyYMARZQAQMVYCKC-OEMFJLHTSA-N
SMILESCC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CCOC1)S(=O)(=O)C1=CC=C(N)C=C1
さらに表示
Appearance (Form)Powder
Appearance (Color)White to beige
Infrared spectrumConforms
HPLC>=97.5 %
Titration with HClO496.0 to 104.0 % (on dry substance)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only