(+)-trans-Taxifolin
Thermo Scientific Chemicals

(+)-trans-Taxifolin

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number 467031000
Price (USD)
236.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
236.00
Each
Chemical Identifiers
CAS480-18-2
IUPAC Name(2R,3R)-2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC15H12O7
InChI KeyCXQWRCVTCMQVQX-LSDHHAIUSA-N
SMILESO[C@@H]1[C@H](OC2=CC(O)=CC(O)=C2C1=O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<3.0 %
Specific optical rotation+19.0° to +23.0° (20°C, 589 nm) (C=1,MeOH)
HPLC>=98.0 %
Appearance (Color)White to yellow to light brown
Appearance (Form)powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only