Fosetyl-aluminum
Thermo Scientific Chemicals

Fosetyl-aluminum

CAS: 39148-24-8 | C6H15AlO9P3, C6H18AlO9P3 | 351.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 467280250
Price (USD)
82.10
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
82.10
Each
Chemical Identifiers
CAS39148-24-8
IUPAC Namealuminium(3+) tris((phosphooxy)ethane)
Molecular FormulaC6H15AlO9P3
InChI KeyOIPMQULDKWSNGX-UHFFFAOYSA-N
SMILES[Al+3].CCOP(=O)=O.CCOP(=O)=O.CCOP(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only