CD39 Monoclonal Antibody (eBioA1 (A1)), APC-eFluor™ 780, eBioscience™ - FAQs
CD39 Monoclonal Antibody (eBioA1 (A1)), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD39 Monoclonal Antibody (eBioA1 (A1)), APC-eFluor™ 780, eBioscience™ - FAQs (47-0399-42, 47-0399-41)