CD4 Monoclonal Antibody (GK1.5), eFluor™ 450, eBioscience™ - FAQs
CD4 Monoclonal Antibody (GK1.5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (GK1.5), eFluor™ 450, eBioscience™ - FAQs (48-0041-80, 48-0041-82)