IL-6 Monoclonal Antibody (MP5-20F3), eFluor™ 450, eBioscience™ - FAQs
IL-6 Monoclonal Antibody (MP5-20F3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-6 Monoclonal Antibody (MP5-20F3), eFluor™ 450, eBioscience™ - FAQs (48-7061-82, 48-7061-80)