IL-12/IL-23 p40 Monoclonal Antibody (eBioHP40 (HP40, HP-40)), eFluor™ 450, eBioscience™ - FAQs
IL-12/IL-23 p40 Monoclonal Antibody (eBioHP40 (HP40, HP-40)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-12/IL-23 p40 Monoclonal Antibody (eBioHP40 (HP40, HP-40)), eFluor™ 450, eBioscience™ - FAQs (48-7235-41, 48-7235-42)