Phospho-STAT3 (Tyr705) Monoclonal Antibody (LUVNKLA), eFluor™ 450, eBioscience™ - FAQs
Phospho-STAT3 (Tyr705) Monoclonal Antibody (LUVNKLA), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Phospho-STAT3 (Tyr705) Monoclonal Antibody (LUVNKLA), eFluor™ 450, eBioscience™ - FAQs (48-9033-41, 48-9033-42)