Cytomat™ 2 Selector Automated Shaking Incubator
Thermo Scientific™

Cytomat™ 2 Selector Automated Shaking Incubator

Simplify your biotherapeutics workflows with the Thermo Scientific™ Cytomat™ 2 Selector Automated Shaking Incubator, featuring large amplitude shaking that overcomes current cell growth limitations.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical Requirements
51035243230V, 50 Hz
51035241100V, 50 Hz
51035242120V, 60 Hz
Catalog number 51035243
Price (USD)
-
Electrical Requirements:
230V, 50 Hz

The Thermo Scientific™ Cytomat™ 2 Selector Automated Shaking Incubator is the first automated incubator for biologics with large amplitude shaking and overcomes current cell growth limitations from low volume form factor to culture vessels (SBS). It enables you to simply copy your manual biologics shaking process inside an automated incubator with a 25 mm shaking diameter and biologically relevant speeds (100-350 rpm). As the newest incubator in the Cytomat 2 C-LiN Series, the Cytomat 2 Selector Incubator comes with the same state-of-the-art features as the classic Cytomat 2 C-LiN Series, providing optimal conditions for cell growth with:

  • Stable temperature
  • Consistent relative humidity
  • CO2 regulation
  • Decontamination routines

The Cytomat 2 Selector Automated Shaking Incubator brings the latest incubation technology to the Cytomat 2 C-LiN Automated Incubator Series with advanced features for biotherapeutics workflows.

  • Designed to accelerate your biologics with stable cell line development.
  • Scale up and simplify your workflow with the ability to automate large volumes with the 25 mm shaking diameter.
  • Set different shaking speeds up to 350 rpm, using two stackers with individual shaking controls.
  • Consistent swirling for gas transfer and cell culture media mixing reduces experiment variation to ease cell selection.
  • Remove risk of contamination in-between runs with the built-in ContraCon™ decontamination routine.
    No removing of hardware required.

Get repeatable results for maximizing your throughput/cell line selection and minimize your variances with the following features:

  • No plate movement with active plate clamping mechanism during shaking
  • Get same results independent of stacker load or plate position
  • No disruption of shaking process during plate handling
  • Ability to unload plate while second stacker is still shaking
  • Device vibration reduction with unique stand design
Specifications
Access TimeMean Access Time
Certifications/ComplianceCE Marked
Depth (English)20.1 in.
Depth (Metric)511 mm
Electrical Requirements230V, 50 Hz
FeaturesShaking incubation, Advanced Orbital Shaking Module with 25 mm shaking diameter, and up to 350 rpm with active clamping
For Use With (Application)Biologics; Cell Line development and cell culture applications requiring 25 mm shaking diameter
Height (English)71.7 in.
Height (Metric)1821 mm
HoldsDepends on Advanced Orbital Shaking (AOS) Module (required); Pitch 52: 16 plates (total); Pitch 95: 8 plates (total); other pitch heights available upon request
ModelCytomat 2 Selector
TypeAutomated Incubator
Weight (Metric)293 kg
Width (English)22.5 in.
Width (Metric)572 mm
Chamber MaterialStainless Steel
Relative Humidity<85%
Temperature Range (Metric)15°C to 50°C
Unit SizeEach