Kanamycin Sulfate
Kanamycin Sulfate
Kanamycin Sulfate
Thermo Scientific Chemicals

Kanamycin Sulfate

CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol
Have Questions?
Catalog NumberQuantity
6112900505 g
Catalog number 611290050
Price (USD)
123.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
123.00
Each
Chemical Identifiers
CAS25389-94-0
IUPAC Name(2R,3S,4S,5R,6R)-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-3-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}oxane-3,4,5-triol; sulfuric acid
Molecular FormulaC18H38N4O15S
InChI KeyOOYGSFOGFJDDHP-KMCOLRRFSA-N
SMILESOS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Identificationpasses test
pH6.5 to 8.5 (1 % solution)
Additional infochromatographic purity: passes test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only