Vanquish™ HPLC and UHPLC System Accessories
Thermo Scientific™
Vanquish™ HPLC and UHPLC System Accessories
Maximize your liquid chromatography (LC) performance with system accessories best suited for the Thermo Scientific™ Vanquish™ HPLC and UHPLC systems.
製品番号(カタログ番号)内容使用対象(アプリケーション)製品ライン
6261.1130Viper Guard ConnectorVanquish Binary Pump N or Metering Device, Viper TQ in Viper only ValvesViper, nano Viper, Viper TQ
6268.0130PEEK Mesh HPLC Solvent FiltersEluent filteringVanquish HPLC and UHPLC Systems
6036.9003Rear Seal and Needle Wash Tubing Labels for Vanquish SystemsLabeling rear seal and needle wash tubing in Vanquish UHPLC SystemsVanquish HPLC and UHPLC Systems
製品番号(カタログ番号) 6261.1130
価格(JPY)
-
内容:
Viper Guard Connector
使用対象(アプリケーション):
Vanquish Binary Pump N or Metering Device, Viper TQ in Viper only Valves
製品ライン:
Viper, nano Viper, Viper TQ
Extend the compatibility and lifespan of nanoViper™ Fignertight Fitting Systems and Viper™ TQ UHPLC Fittings with the Viper Guard Connector. Add the Thermo Scientific™ PEEK Mesh HPLC solvent filter and Rear Seal and Needle Wash Tubing Labels for Vanquish LC Systems for enhance user-friendliness.
Optimizing the chromatographic performance and user-friendliness with accessories suited best for Vanquish HPLC and UHPLC systems.
仕様
使用対象(アプリケーション)Vanquish Binary Pump N or Metering Device, Viper TQ in Viper only Valves
内容Viper Guard Connector
材料MP35N, PEEK
最大圧力1517 bar
使用対象 (装置)Vanquish Neo UHPLC Systems, Viper TQ, Viper and nanoViper capillaries
製品ラインViper, nano Viper, Viper TQ
タイプAdapter
Unit SizeEach