Nalgene™ Polyolefin Right-To-Know Custom Labeling System
Nalgene™ Polyolefin Right-To-Know Custom Labeling System
Nalgene™ Polyolefin Right-To-Know Custom Labeling System
Nalgene™ Polyolefin Right-To-Know Custom Labeling System
Thermo Scientific™

Nalgene™ Polyolefin Right-To-Know Custom Labeling System

Create labels that identify a chemical and its properties using the Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System.
製品番号(カタログ番号)高さ(メートル法)幅(メートル法)
6316-1000PK
または、製品番号6316-1000
76.2 mm127 mm
製品番号(カタログ番号) 6316-1000PK
または、製品番号6316-1000
価格(JPY)
24,780
Pack of 25
高さ(メートル法):
76.2 mm
幅(メートル法):
127 mm

Create labels that identify a chemical and its properties using the Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System.

Features:
  • Overlay provided to protect label from chemical contact
  • Complete instructions included with each package
  • Hazard symbol stickers are GHS-compliant
Note: Refer to the appropriate Material Safety Data Sheet (MSDS) or other reference material to verify the correct codes required for accurate and safe use.

仕様
概要Custom Right-To-Know Label System
高さ(ヤードポンド法)3 in.
高さ(メートル法)76.2 mm
箱あたりの入数150
パックあたり入数25
幅(メートル法)127 mm
色Assorted
フォーマットLabel
材料Polyolefin
幅(ヤードポンド法)5 in.
Unit SizePack of 25