CD4 Monoclonal Antibody (RM4-5), eFluor™ 506, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RM4-5), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RM4-5), eFluor™ 506, eBioscience™ - FAQs (69-0042-82, 69-0042-80)