CaptureSelect™ Biotin Anti-AdV5 Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-AdV5 Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-AdV5 Conjugate consists of a 13-kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to hexon capsomeres of Adenovirus serotype 5 (AdV5) particles as well as free hexon proteins thereof.
Catalog NumberQuantity
7103532100100 μg
7103532500500 μg
Catalog number 7103532100
Price (JPY)
85,200
Each
Quantity:
100 μg
CaptureSelect Biotin Anti-AdV5 Conjugate consists of a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to hexon capsomeres of Adenovirus serotype 5 (AdV5) particles as well as free hexon proteins thereof. Possible cross-reactivity towards hexon proteins of other Adenovirus serotypes has not been tested.
The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize AdV5 virus particles.
Applications for CaptureSelect Biotin Anti-AdV5 Conjugate include ELISA, immunoprecipitation with streptavidin-coupled Dynabeads, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeBiotin & Analogs
Quantity100 μg
Shipping ConditionDry Ice
FormLiquid
Product LineCaptureSelect
TypeConjugate
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).