Nalgene™ Syringe Filters, Non-sterile
Nalgene™ Syringe Filters, Non-sterile
Nalgene™ Syringe Filters, Non-sterile
Nalgene™ Syringe Filters, Non-sterile
Thermo Scientific™

Nalgene™ Syringe Filters, Non-sterile

Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
製品番号(カタログ番号)孔径直径(メートル法)材料(膜)
724-2020PK
または、製品番号724-2020
0.2 μm25 mmSFCA
721-13200.2 μm13 mmPTFE
721-13450.45 μm13 mmPTFE
722-20001.1 μm25 mmGlass Fiber
724-2045PK
または、製品番号724-2045
0.45 μm25 mmSFCA
724-2080PK
または、製品番号724-2080
0.8 μm25 mmCellulose Acetate
727-2020PK
または、製品番号727-2020
0.2 μm25 mmNylon
727-2045PK
または、製品番号727-2045
0.45 μm25 mmNylon
728-2020PK
または、製品番号728-2020
0.2 μm25 mmPTFE
728-2045PK
または、製品番号728-2045
0.45 μm25 mmPTFE
176-0020PK
または、製品番号176-0020
0.2μm4mmナイロン
176-0045PK
または、製品番号176-0045
0.45μm4mmナイロン
製品番号(カタログ番号) 724-2020PK
または、製品番号724-2020
価格(JPY)
16,460
キャンペーン価格
Ends: 25-Sep-2026
29,140
割引額 12,680 (44%)
Pack of 100
孔径:
0.2 μm
直径(メートル法):
25 mm
材料(膜):
SFCA

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

仕様
直径(メートル法)25 mm
直径(メートル法)膜25 mm
使用対象(アプリケーション)Filtration of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids
グリーン機能Fewer resources used and less waste
ハウジング材質Polypropylene (Housing)
製品ラインNalgene
タイプSyringe Filter
容量(メートル法)10 to 100 mL (Sample)
材料(膜)SFCA
孔径0.2 μm
無菌性Non-sterile
Unit SizePack of 100