Adapters for Oval Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Thermo Scientific™
Adapters for Oval Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Choice of 4 adapters to accommodate a range of blood bags in oval single blood bag buckets for Thermo Scientific™ HAEMAFlex™ 6/8 rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
750038416/8 x 450 mL Single or Double Blood Bag
750038386/8 x 450 mL Double or Triple Blood Bag
750038396/8 x 250 mL Platelet-rich Plasma or Buffy Coat Bag
750038376/8 x 550 mL Quad or Quint Blood Bag
Catalog number 75003841
Price (USD)
-
Capacity:
6/8 x 450 mL Single or Double Blood Bag
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
Select adapters for 450–550mL Quad or Quint blood bags, 400–450mL Double or Triple blood bags, 250mL Platelet-rich Plasma or Buffy Coat bags or 400–450mL Single or Double blood bags.
Specifications
For Use With (Equipment)Oval single blood bag buckets
No. per PackSet of 2
Capacity6/8 x 450 mL Single or Double Blood Bag
TypeBlood Bag Adapter
Unit SizeEach