Orion™ 8030cX Silica Analyzer
Thermo Scientific™

Orion™ 8030cX Silica Analyzer

Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica.
製品番号(カタログ番号)タイプ数量
8030CLCleaning Solution1 Ea.
7070FLTSample Prefilter1 Ea.
803002Silica Standard Solution20 μg/L
803020Silica Standard Solution200 μg/L
803030Silica Standard Solution1000 μg/L
803040Silica Standard Solution4000 μg/L
8030CXSilica Analyzer1 Ea.
8030MKMaintenance Kit1 Ea.
8010PCMProfibus Card1 Ea.
8030REXSilica Reagent Kit1 Ea.
製品番号(カタログ番号) 8030CL
価格(JPY)
-
タイプ:
Cleaning Solution
数量:
1 Ea.
Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica. Powered by advanced colorimetric technology, the Orion 8030cX delivers premium precision, greater productivity and enhanced durability. Supporting ultrapure and process water applications, the Orion 8030cX conveniently delivers precise, reliable measurement you can depend on. Unleash the possibilities and conquer the freedom of fast, confident decision making.
Confident Results
  • Low Detection Limit (0.5 ppb)
  • High Accuracy (≤1 ppb or 1% for range 0-500 ppb)
  • Patented Fluid Delivery & Quantification System
  • Reduced Phosphate Interferences

 Low Cost Ownership
  • 100 Days of Continuous Run Time (w/ 2L reagent and 15-minute measurement frequency)
  • Low Reagent Consumption (200 μL per measurement)
  • Separate Chemical Waste Collection (decreased disposal expenses)

 Easy to Use
  • Automated Calibration
  • Automated Validation & Verification
  • Color Touchscreen with Intuitive Interface

 Maximum Uptime
  • Automated Cleaning
  • Scheduled Measurements
  • Alert Notification System
  • Alarm System w/ Troubleshooting Guide
  • IP65 Certified (splash/ dust proof)
Markets
Silica monitoring for ultrapure water, demineralized water, deionized water, boiler water and steam condensate in:
  • Power Generation
  • Oil & Gas
  • Chemical Production
  • Semiconductor Manufacturing
  • Pulp & Paper


Applications

Monitoring and controlling silica levels in ultrapure and industrial process water to:
  • Prevent buildup on turbine blades, heat exchangers, boilers and more
  • Reduce costly and time-consuming cleaning procedures
  • Identify reverse osmosis (RO) breakdown and saturation of ion exchange resins
  • Maximize thermal efficiencies of steam passageways
  • Safeguard the health of critical plant assets by reducing potential for turbine vibration
仕様
概要The Thermo Scientific™ Orion™ 8030cX Silica Cleaning Solution, includes one (1) 1L cleaning solution to be used together with 8030REX for analysis.
使用対象 (装置)Orion 8030cX Silica Analyzer
数量1 Ea.
タイプCleaning Solution
Unit SizeEach