Product Image
Thermo Scientific™
Nalgene™ Polypropylene, Carboy with Handle
For Use With: For use with large batches of culture media, vaccine product production, bulk media and chemical storage; Autoclavable; Closure Material: Polypropylene; Color: White
製品番号(カタログ番号)数量
8250-0020PK
または、製品番号8250-0020
Case of 6 or Pack of 1
製品番号(カタログ番号) 8250-0020PK
または、製品番号8250-0020
価格(JPY)
17,000
Pack of 1
数量:
Case of 6 or Pack of 1
Choose Thermo Scientific™ Nalgene™ Autoclavable Carboys for large batches of culture media, vaccine product production, bulk media and chemical storage. These carboys feature a large mouth for easy filling/dispensing, and are made of natural polypropylene with white 83B polypropylene closure and TPE (thermoplastic elastomer) gasket.
Customization options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.
Key Features:
  • Convenient shoulder handles allow easy carrying and pouring, even with gloved hands
  • Available in 10, 20, and 50L sizes for a variety of biopharmaceutical applications
  • Graduated to contain in 1 gal. and 5L increments for ease of use
  • Nalgene Top Works Fluid Transfer Systems (Cat. No. 2135) permits convenient liquid aseptic transfer
  • Autoclavable/Graduated/Leakproof†
Ordering Information:
  • Additional closure options are Nalgene Quick Filling/Venting Closures (Cat. No. 2158) or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149)
  • Custom Tubing Sets are available-please refer to Thermo Scientific Custom Tubing Set Configurator at www.thermoscientific.com/tubingconfigurator.com
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
仕様
容量(ヤードポンド法)2.6ガロン
クロージャー材質ポリプロピレン
クロージャーサイズ83B
概要Polypropylene Carboy w/Handles (Manufactured in our state-of-the-art Suzhou, China facility)
直径(メートル法)外径250 mm
使用対象(アプリケーション)For use with large batches of culture media, vaccine product production, bulk media and chemical storage
高さ(メートル法)クロージャー付き389 mm
内容Handles
箱あたりの入数6/Cs.
パックあたり入数1 Ea.
数量Case of 6 or Pack of 1
容量(ヤードポンド法)満杯3 gal.
容量(メートル法)満杯12 L
オートクレーブ可能Autoclavable
容量(メートル法)10 L
White
材料Polypropylene
製品タイプCarboy with Handle
Unit SizePack of 1