MarqMetrix Performance BallProbe Sampling Optic
MarqMetrix Performance BallProbe Sampling Optic
MarqMetrix Performance BallProbe Sampling Optic
MarqMetrix Performance BallProbe Sampling Optic
Thermo Scientific™

MarqMetrix Performance BallProbe Sampling Optic

The Thermo Scientific™ MarqMetrix™ Performance BallProbe™ is built for lab and industrial use, offering sensitive TouchRaman Immersion measurements in moderate chemical environments across various sample types.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de productoDiseño óptico
840-401300BallProbeBola de zafiro de grado UV de 8 mm de diámetro
840-401700BallProbeBola de zafiro de grado UV de 6 mm de diámetro
840-401800BallProbeBola de zafiro de grado UV de 3 mm de diámetro
840-399700BallProbe con cabezal de fibraBola de zafiro de grado UV de 8 mm de diámetro
840-400200BallProbe con cabezal de fibraBola de zafiro de grado UV de 6 mm de diámetro
840-400300BallProbe con cabezal de fibraBola de zafiro de grado UV de 3 mm de diámetro
Número de catálogo 840-401300
Precio (CLP)
-
Tipo de producto:
BallProbe
Diseño óptico:
Bola de zafiro de grado UV de 8 mm de diámetro
The Thermo Scientific™MarqMetrix™ Performance BallProbe™ Sampling Optic is designed to withstand moderate chemical environments in laboratory and industrial settings. The standard 0.5 in. diameter MarqMetrix Performance BallProbe Sampling Optic utilizes our largest spherical sapphire ball lens and 316 L stainless steel, enabling highly sensitive TouchRaman Immersion Technology measurements across a wide range of sample types. The MarqMetrix Performance BallProbe Sampling Optic has three standard diameters and chassis length options and is also available in a bundle with an interchangeable fiber head.

Features and benefits

  • Accurate and repeatable measurements
  • Samples liquids, solids, slurries, powders, and heterogeneous mixtures
  • Wide range of chemical compatibility and resistance to high-temperatures
  • Ease of use eliminates user-to-user variation

Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Food and beverage processing
  • Oil and gas processing
  • Polymer and plastic manufacturing
  • Chemical and hazardous applications

Operating conditions

  • Suitable for intermittent exposure to most dilute and concentrated acids (hot and cold), bases, and most organic solvents, including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane, and acetonitrile
  • Avoid exposure to aqua regia, heated formic acid, mixed acid, TFAA
Especificaciones
Compatible Wavelengths500 to 1100 nm
Descripción1/2 in. Performance BallProbe (200 mm) Accepts a collimated laser beam up to 7.1 mm (0.275 in.) in diameter
Para utilizar con (equipo)MarqMetrix All-In-One Process Raman Analyzer
MaterialHigh-temperature epoxy (wetted), Stainless Steel (316L)
Diseño ópticoBola de zafiro de grado UV de 8 mm de diámetro
Pressured Design Condition6000 psi (413 bar)
Longitud de la sonda8.5 in. (200 mm)
Probe Outside Diameter0.050 in. (12.7 mm)
Intervalo de temperatura (métrico)- 20°C to 150°C
PantallaNo Display
Tipo de productoBallProbe
Unit SizeEach