Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Imagem ilustrativa. O produto pode variar
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Thermo Scientific™

Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing

Ensure consistently reliable and accurate fluid transfers. This durable high-purity tubing is ideal for pharmaceutical, laboratory and bioprocess manufacturing.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoDiameter (English) InnerThickness (English)
8600-01303/8 in.3/32 in.
8600-00201/16 in.1/32 in.
8600-00301/8 in.1/16 in.
8600-00403/16 in.1/16 in.
8600-00503/16 in.3/32 in.
8600-00601/4 in.1/16 in.
8600-00701/4 in.3/32 in.
8600-00801/4 in.1/8 in.
8600-01005/16 in.3/32 in.
8600-01203/8 in.1/16 in.
8600-01403/8 in.1/8 in.
8600-01901/2 in.3/32 in.
8600-02001/2 in.1/8 in.
8600-02605/8 in.1/8 in.
8600-03103/4 in.1/8 in.
Número do catálogo 8600-0130
Preço (BRL)
-
Diameter (English) Inner:
3/8 in.
Thickness (English):
3/32 in.

Ensure consistently reliable and accurate fluid transfers with Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing for Peristaltic Pumpsis.This flexible, durable high-purity translucent tubing is designed for a variety of pump and transfer applications, including pharmaceutical, laboratory and bioprocess manufacturing.

  • Tasteless, odorless, non-cytotoxic and non-hemolytic
  • Contains no phthalate plasticizers or peroxides that can leach out into transported fluid
  • Low binding suface to minimize absorption of proteins and food products
  • Autoclavable; also can be sterilized by gamma radiation, EtO and chemical procedures
  • Durometer hardness: Shore A, 50

Ordering Information:  Double-bagged in case of 50 ft. (15.2m)

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Especificações
Certifications/ComplianceMeets USP<87> Cytotoxicity (Agar Diffusion and MEM Elution methods); USP<88> Class VI; USP<661>Physicochemical Tests; ISO10993-3 Bacterial Mutagenicity – AmesAssay; ISO10993-4 Hemolysis Direct Contact, EP 3.1.9 Silicone Elastomer f
Flexibility4 x OD bend radius
Hardness50 (Shore A)
Maximum Pressure12 psig at 73°F (23°C)
Thickness (English)3/32 in.
Thickness (Metric)2.4 mm
AutoclavableAutoclavable
ColorTranslucent
Diameter (English) Inner3/8 in.
Diameter (English) Outer9/16 in.
Diameter (Metric) Inner9.5 mm
Diameter (Metric) Outer14 mm
MaterialPlatinum-cured Silicone
Overall Length (English)50 ft.
Product TypeFlexible Tubing
Unit SizeCase of 50