Product Image
Invitrogen™
eBioscience™ Intracellular Fixation & Permeabilization Buffer (plus Brefeldin A)
The eBioscience Fixation and Permeabilization Kit is designed for use in preparation of living cells for intracellular staining and flowRead more
Have Questions?
Catalog NumberQuantity
88-8823-881 kit
Catalog number 88-8823-88
Price (USD)
340.00
Each
Quantity:
1 kit
Price (USD)
340.00
Each
The eBioscience Fixation and Permeabilization Kit is designed for use in preparation of living cells for intracellular staining and flow cytometric analysis. A protein transport inhibitor (Brefeldin A) is included for treatment of cells during culture – to cause accumulation of cytokine protein at the Golgi and enhance resulting staining. A fixation buffer is used to cross link, stabilize, and fix the cell membrane, while the permeabilization buffer is used to reversibly permeabilize the cell membrane so the staining antibodies can enter the cell effectively.
Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Reagent TypeFixation Reagent, Permeabilization Reagent
Product LineeBioscience
Product TypeFixation/Permeabilization Buffer
Unit SizeEach