Product Image
Invitrogen™
MagniSort™ Human T cell Enrichment Kit
The MagniSort™ Human T cell Enrichment Kit is designed for the magnetic separation of T cells by negative selection. It深入閱讀
Have Questions?
產品號碼Quantity
8804-6810-74200 tests
產品號碼 8804-6810-74
價格 (HKD)
6,403.00
線上優惠
Ends: 31-Dec-2026
9,148.00
您節省 2,745.00 (30%)
Each
Quantity:
200 tests
價格 (HKD)
6,403.00
線上優惠
Ends: 31-Dec-2026
9,148.00
您節省 2,745.00 (30%)
Each
The MagniSort™ Human T cell Enrichment Kit is designed for the magnetic separation of T cells by negative selection. It has been optimized for the isolation of human T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave T cells untouched and free in solution.
The MagniSort™ Human T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
規格
Cell TypeT Cells
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypeCell Enrichment Kit
Unit SizeEach