Product Image
Invitrogen™
MagniSort™ Human CD8 Naïve T cell Enrichment Kit
The MagniSort™ Human CD8 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD8 naïve T cellsRead more
Have Questions?
Catalog NumberQuantity
8804-6816-74200 tests
Catalog number 8804-6816-74
Price (TWD)
***
Quantity:
200 tests
Price (TWD)
***
The MagniSort™ Human CD8 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD8 naïve T cells by negative selection. It has been optimized for the isolation of human CD8 naïve T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 naïve T cells untouched and free in solution.
The MagniSort™ Human CD8 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD4 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RO Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (Naïve, CD4)
No. of Tests200 tests
Quantity200 tests
Target SpeciesHuman
Product TypeCell Enrichment Kit
Unit SizeEach