GeneChip™ Rat Genome 230 2.0 Assay
Actual product may vary
Applied Biosystems™

GeneChip™ Rat Genome 230 2.0 Assay

The GeneChip™ Rat Genome 230 2.0 Assay is a powerful tool for toxicology, neurobiology, and other applications using rat asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90249330 arrays
90249210 arrays
Catalog number 902493
Price (USD)
-
Number of Arrays:
30 arrays
The GeneChip™ Rat Genome 230 2.0 Assay is a powerful tool for toxicology, neurobiology, and other applications using rat as a model organism.

• Provides comprehensive coverage of the transcribed rat genome on a single array
• Comprised of more than 31,000 probe sets, analyzing over 30,000 transcripts and variants from over 28,000 well-substantiated rat genes
• Sophisticated bioinformatics tools available through the NetAffx™ Analysis Center to rapidly obtain biologically meaningful results
• Publicly available drafts of the rat genome and leading public rat databases were used to refine sequences and provide a higher quality of data output

Assay Profile
All probe sets represented on the GeneChip Rat Expression Set 230 are included on the GeneChip Rat Genome 230 2.0 Assay. Sequences used in the design of the GeneChip Rat Genome 230 2.0 Assay were selected from GenBank™, dbEST, and RefSeq. The sequence clusters were created from the UniGene database (Build 99, June 2002) and then refined by analysis and comparison with the publicly available draft assembly of the rat genome from the Baylor College of Medicine Human Genome Sequencing Center (June 2002).

The GeneChip Rat Genome 230 2.0 Assay includes representation of the RefSeq database sequences and probe sets related to sequences and refined EST clusters previously represented on the GeneChip Rat Genome U34 Set.

Oligonucleotide probes complementary to each corresponding sequence are synthesized in situ on the arrays. Eleven pairs of oligonucleotide probes are used to measure the level of transcription of each sequence represented on the GeneChip Rat Genome 230 2.0 Assay.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Product LineGeneChip
Quantity30 reactions
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays30 arrays
SpeciesRat
Unit SizeEach

Frequently asked questions (FAQs)

What reagent kit should I use with my array?

Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Is it possible to compare different array types (e.g., Rat 230A and Rat 230 2.0) in GeneChip Command Console (GCC)?

GeneChip Command Console cannot generate a comparison analysis between different array types.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How long does the scan of a Rat 230 2.0 Array take?

Scanning a single Rat 230 2.0 array takes approximately 9 minutes. This is about 25% less time than is required to scan the Rat 230A and Rat 230B arrays combined.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How much memory is needed to store the data from a single Rat 230 2.0 Array?

A single Rat 230 2.0 array generates approximately 92 MB of data.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Where can I find the high-resolution scanning patch for the Rat 230 2.0 Array?

Please contact Technical Support to obtain the high-resolutiion scanning patch.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.