AcroMetrix™ Transplant Virus Panels (RUO)
Product Image
Thermo Scientific™

AcroMetrix™ Transplant Virus Panels (RUO)

Assess the analytical performance of molecular test procedures for transplant viruses with these panels, which are for Research Use Only.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionPackagingLevel
954525AcroMetrix HSV-2 Plasma Panel5 x 0.5mL100, 1000, 10000, 100000, 1000000 copies/mL
954530AcroMetrix VZV Plasma Panel5 x 0.5mL100, 1000, 10000, 100000, 1000000 copies/mL
942017AAcroMetrix EBV Plasma Panel5 x 0.5mL100, 1000, 10000, 100000, 1000000 IU/mL
942016AcroMetrix CMVtc Panel5 x 0.8mL300, 3000, 30000, 300000, 3000000 IU/mL
954520AcroMetrix HSV-1 Plasma Panel5 x 0.5mL100, 1000, 10000, 100000, 1000000 copies/mL
954535AcroMetrix Adenovirus Plasma Panel5 x 0.5mL1000, 10000, 100000, 1000000, 10000000 copies/mL
950360AcroMetrix BKV Panel5 x 0.5mL500, 5000, 50000, 500000, 5000000 copies/mL
Catalog number 954525
Price (USD)
-
Request A Quote
Description:
AcroMetrix HSV-2 Plasma Panel
Packaging:
5 x 0.5mL
Level:
100, 1000, 10000, 100000, 1000000 copies/mL
Assess the analytical performance of molecular test procedures, including the nucleic acid extraction step, for the quantitative and qualitative determination of viruses related to transplant disease with the Thermo Scientific™ AcroMetrix™ Transplant Virus Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits, and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionAcroMetrix HSV-2 Plasma Panel
Level100, 1000, 10000, 100000, 1000000 copies/mL
Packaging5 x 0.5mL
Quantity5 x 0.5 mL
TypePlasma Panel
Unit SizeEach