Thermo Scientific Chemicals

o-Tolualdehyde, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals

Catalog number: A10172.14
25 g, Each
Thermo Scientific Chemicals

o-Tolualdehyde, 98%, stab. with 0.1% hydroquinone, Thermo Scientific Chemicals

Catalog number: A10172.14
25 g, Each
Quantity
Catalog number: A10172.14
also known as A10172-14
Price (USD)
Price: 62.20
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
529-20-4
IUPAC Name
2-methylbenzaldehyde
Molecular Formula
C8H8O
InChI Key
BTFQKIATRPGRBS-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1C=O
Appearance (Color)
Clear colorless to yellow or pale orange
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.5460-1.5550 @ 20°C

Description

o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.

Solubility
Soluble in carbon tetrachloride, ethanol, ethyl ether, benzene. Not miscible in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text