3,5-Dichloro-2,4,6-trifluoropyridine, 97+%
3,5-Dichloro-2,4,6-trifluoropyridine, 97+%
3,5-Dichloro-2,4,6-trifluoropyridine, 97+%
3,5-Dichloro-2,4,6-trifluoropyridine, 97+%
Thermo Scientific Chemicals

3,5-Dichloro-2,4,6-trifluoropyridine, 97+%

CAS: 1737-93-5 | C5Cl2F3N | 201.957 g/mol
製品番号(カタログ番号) A10205.22
または、製品番号A10205-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1737-93-5
IUPAC Name3,5-dichloro-2,4,6-trifluoropyridine
Molecular FormulaC5Cl2F3N
InChI KeyPKSORSNCSXBXOT-UHFFFAOYSA-N
SMILESFC1=NC(F)=C(Cl)C(F)=C1Cl
さらに表示
Appearance (Color)Clear colorless to pale yellow or white to pale yellow
Assay (GC)≥97.0%
FormLiquid as melt or crystalline powder and/or fused solid
Melting Point (clear melt)22-28°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only