4-Amino-3-hydroxybenzoic acid, 98%
4-Amino-3-hydroxybenzoic acid, 98%
4-Amino-3-hydroxybenzoic acid, 98%
4-Amino-3-hydroxybenzoic acid, 98%
Thermo Scientific Chemicals

4-Amino-3-hydroxybenzoic acid, 98%

CAS: 2374-03-0 | C7H7NO3 | 153.137 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A10207.22
または、製品番号A10207-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2374-03-0
IUPAC Name4-amino-3-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyNFPYJDZQOKCYIE-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1O)C(O)=O
さらに表示
Appearance (Color)Dark cream to cream to brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. specification)
Assay (Silylated GC)≥97.5% (non-U.S. specification)
Assay from Suppliers CofA≥97.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
さらに表示
4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydroxybenzoic acid is used in the preparation of various pharmaceutical compounds such as sphingosine kinase inhibitors.

Solubility
Soluble in chloroform, and aqeous ethanol, water (Partly miscible)

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
RUO – Research Use Only
Chika Orii; Shinji Takenaka; Shuichiro Murakami; and Kenji Aoki . A novel coupled enzyme assay reveals an enzyme responsible for the deamination of a chemically unstable intermediate in the metabolic pathway of 4-amino-3-hydroxybenzoic acid in Bordetella sp. strain 10d. Eur. J. Biochem. 2004, 271,(15), 3248-3254.