1-Adamantanol, 99%
1-Adamantanol, 99%
1-Adamantanol, 99%
Thermo Scientific Chemicals

1-Adamantanol, 99%

CAS: 768-95-6 | C10H16O | 152.237 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A10209.18
または、製品番号A10209-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS768-95-6
IUPAC Nameadamantan-1-ol
Molecular FormulaC10H16O
InChI KeyVLLNJDMHDJRNFK-UHFFFAOYSA-N
SMILESOC12CC3CC(CC(C3)C1)C2
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanol was used in the preparation of 1,3-adamantanediol. It is used in the manufacture of synthetic adamantane derivatives and adapalene.

Solubility
Soluble in ethanol , methanol, and diethyl ether. Partly miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
RUO – Research Use Only

General References:

  1. Koichi Mitsukura; Hiromu Sakamoto; Haruka Kubo; Toyokazu Yoshida; Toru Nagasawa. Bioconversion of 1-adamantanol to 1,3-adamantanediol usingStreptomycessp. SA8 oxidation system. Journal of Bioscience and Bioengineering. 2010, 109 (6), 550-553.
  2. Motowo Yamaguchi; Hiroyuki Kousaka; Shinichi Izawa; Yoshiki Ichii; Takashi Kumano; Dai Masui; Takamichi Yamagishi. Syntheses, characterization, and catalytic ability in alkane oxygenation of chloro(dimethyl sulfoxide)ruthenium(II) complexes with tris(2-pyridylmethyl)amine and its derivatives. Inorganic Chemistry. 2006, 45 (20), 8342-8354.
  3. Carboxyl groups of amino acids have been protected as 1-adamantyl esters via the amino acid tosylate and dimethyl sulfite. The group is readily cleaved with TFA under mild conditions: Synthesis, 1355 (1996). Has also been applied in blocking the ß-carboxyl group of aspartic acid in peptide synthesis; the 1-adamantyl ester is prepared using DCC-DMAP: J. Chem. Soc., Perkin 1, 2129 (1988). See also Appendix 6.
  4. Thiol groups may also be protected by formation of the thioether in TFA. Cleavage is by means of Hg(II) salts: Chem. Pharm. Bull., 26, 1576 (1978).
  5. Reaction with Hydrogen fluoride pyridine complex, L17117, gives 1-fluoroadamantane, a general method for conversion of sec- and tert-alcohols to fluoroalkanes: Org. Synth. Coll., 6, 628 (1988).