Thermo Scientific Chemicals

Polyethylene powder, low density, 500 micron, Thermo Scientific Chemicals

Catalog number: A10239.0E
2500 g, Each
Thermo Scientific Chemicals

Polyethylene powder, low density, 500 micron, Thermo Scientific Chemicals

Catalog number: A10239.0E
2500 g, Each
Quantity
Catalog number: A10239.0E
also known as A10239-0E
Price (USD)
Price: 526.00
Online price: 447.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9002-88-4
Molecular Formula
(C2H4)n
SMILES
*-CC-*
MDL Number
MFCD00084423
Density @20°C
(Resin): 0.9100-0.9600 g/ml
Form
Powder
Particle Size
approx. 500 micron
Appearance (Color)
White
Comment
Melt flow index: 17.5-23.5 g/10min (190?C/2.16kg)

Description

Polyethylene powder is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for manufacturing various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text