5-Chloro-2-pentanone, 94%
5-Chloro-2-pentanone, 94%
5-Chloro-2-pentanone, 94%
5-Chloro-2-pentanone, 94%
Thermo Scientific Chemicals

5-Chloro-2-pentanone, 94%

CAS: 5891-21-4 | C5H9ClO | 120.576 g/mol
製品番号(カタログ番号) A10269.36
または、製品番号A10269-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5891-21-4
IUPAC Name5-chloropentan-2-one
Molecular FormulaC5H9ClO
InChI KeyXVRIEWDDMODMGA-UHFFFAOYSA-N
SMILESCC(=O)CCCCl
さらに表示
Assay (GC)≥92.0%
Refractive Index1.4355-1.4425 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to yellow to dark brown to brown
Identification (FTIR)Conforms
5-Chloro-2-pentanone used in the preparation of destructible surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-pentanone used in the preparation of destructible surfactants.

Solubility
Soluble in chloroform and methanol.

Notes
keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents
RUO – Research Use Only

General References:

  1. Hideo Kise; Toru Sato; Tsutomu Yasuoka; Manabu Seno; Teruzo Asahara. Synthesis of ocimenol and related compounds from 5-chloro-2-pentanone. J. Org. Chem. 1979, 44, (24), 4454-4456.
  2. David A. Jaeger; Moira R. Frey. Preparation and characterization of destructible surfactants. J. Org. Chem. 1982, 47, (2), 311-315.