Thermo Scientific Chemicals

o-Anisidine, 99%, Thermo Scientific Chemicals

Catalog number: A10288.0B
1000 g, Each
Thermo Scientific Chemicals

o-Anisidine, 99%, Thermo Scientific Chemicals

Catalog number: A10288.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10288.0B
also known as A10288-0B
Price (USD)
Price: 118.00
Your price:
Quantity
-

Chemical Identifiers

CAS
90-04-0
IUPAC Name
2-methoxyaniline
Molecular Formula
C7H9NO
InChI Key
VMPITZXILSNTON-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1N
Appearance (Color)
Clear yellow to red or brown
Assay (GC)
≥98.5%
Refractive Index
1.5735-1.5765 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text