General Description
• L-Methionine is an essential amino acid required for tissue repair and maintenance
• L-Methionine affects absorption of selenium and zinc
• L-Methionine can assist cellular detoxification by chelating lead and mercury
• L-Methionine exhibitis properties associated with anti-aging and can also prevent the buildup of fat in the liver
Application
• L-Methionine is appropriate for use in cell culture and biochemistry laboratory applications
• It is useful for the laboratory study of methionine pathways in the cell relating to aging, degeneration, and metabolism