2,3-Dimethylbenzoic acid, 98%
2,3-Dimethylbenzoic acid, 98%
2,3-Dimethylbenzoic acid, 98%
Thermo Scientific Chemicals

2,3-Dimethylbenzoic acid, 98%

CAS: 603-79-2 | C9H10O2 | 150.177 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A10383.14
または、製品番号A10383-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS603-79-2
IUPAC Name2,3-dimethylbenzoic acid
Molecular FormulaC9H10O2
InChI KeyRIZUCYSQUWMQLX-UHFFFAOYSA-N
SMILESCC1=CC=CC(C(O)=O)=C1C
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)142.5-148.5?C
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only