Azobenzene, 97+%
Azobenzene, 97+%
Azobenzene, 97+%
Thermo Scientific Chemicals

Azobenzene, 97+%

CAS: 103-33-3 | C12H10N2 | 182.23 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A10425.22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS103-33-3
Appearance (Color)Orange to red
Assay (GC)≥97.0%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or chunks
Melting Point (clear melt)62.0-72.0?C
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.

Solubility
Soluble in alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Store at 4°C. Incompatible with strong oxidizing agents. Air and light sensitive.
RUO – Research Use Only
Andreas Archut; Fritz Vögtle; Luisa De Cola; Gianluca Camillo Azzellini; Vincenzo Balzani; P. S. Ramanujam and Rolf H. Berg. Azobenzene-Functionalized Cascade Molecules: Photoswitchable Supramolecular Systems. Chemistry - A European Journal.1998, 4, 699-706.