1-Bromopropane, 99%
1-Bromopropane, 99%
1-Bromopropane, 99%
Thermo Scientific Chemicals

1-Bromopropane, 99%

CAS: 106-94-5 | C3H7Br | 122.993 g/mol
数量:
1000 mL
5000 mL
製品番号(カタログ番号) A10461.0J
または、製品番号A10461-0J
価格(JPY)
-
見積もりを依頼する
数量:
5000 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS106-94-5
IUPAC Name1-bromopropane
Molecular FormulaC3H7Br
InChI KeyCYNYIHKIEHGYOZ-UHFFFAOYSA-N
SMILESCCCBr
さらに表示
Appearance (Color)Clear colorless to pale yellow
CommentMay darken on storage
Identification (FTIR)Conforms (non-U.S. specification)
Water Content (Karl Fischer Titration)≤0.01% (100 ppm) (U.S. specification)
Refractive Index1.4325-1.4360 @ 20°C (non-U.S. specification)
さらに表示
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

Solubility
Miscible with ethanol, water and diethyl ether.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Garner, C. E.; Liang, S.; Yin, L.; Yu, X. Physiologically based pharmacokinetic modeling for 1-bromopropane in F344 rats using gas uptake inhalation experiments. Toxicol. Sci. 2015, 145 (1), 23-36.
  2. Frasch, H. F.; Dotson, G. S.; Barbero, A. M. In vitro Human Epidermal Penetration of 1-Bromopropane. J. Toxicol. Environ. Health A 2011, 74 (19), 1249-1260.